Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADCY7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114.4 | NP_001105.1 | 17 | 2357 | ||
NM_001286057.1 | NP_001272986.1 | 18 | 2405 | |||
XM_011522835.2 | XP_011521137.1 | 18 | 2458 | |||
XM_011522836.2 | XP_011521138.1 | 18 | 2474 | |||
XM_011522837.2 | XP_011521139.1 | 18 | 2403 | |||
XM_011522838.2 | XP_011521140.1 | 18 | 2607 | |||
XM_011522839.2 | XP_011521141.1 | 18 | 2434 | |||
XM_011522840.2 | XP_011521142.1 | 18 | 2441 | |||
XM_011522842.2 | XP_011521144.1 | 18 | 2504 | |||
XM_017022895.1 | XP_016878384.1 | 18 | 2474 | |||
XM_017022896.1 | XP_016878385.1 | 15 | 1620 | |||
XM_017022897.1 | XP_016878386.1 | 15 | 1639 | |||
XM_017022898.1 | XP_016878387.1 | 18 | 2474 | |||
XM_017022899.1 | XP_016878388.1 | 18 | 2474 | |||
GenBank mRNA | AK093706.1 | - | 19 | 2355 | ||
BC039891.1 | - | 15 | 1993 | |||
BC126271.1 | - | 17 | 2169 | 8991987 | ||
D25538.1 | - | 17 | 2354 |