Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANK3 SIRNA Assays ›
Gene Symbol:
ANK3
Gene Name:
ankyrin 3, node of Ranvier (ankyrin G)
Gene Aliases:
ANKYRIN-G, MRT37
Chromosome Location:
Chr.10: 60026298 - 60733526 on Build GRCh38
UniGene ID:
Hs.499725
Species:
Human
siRNA ID
HSS100488
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANK3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001149.3
NP_001140.2
4, 5
996
NM_001204403.1
NP_001191332.1
27, 28
3134
NM_001204404.1
NP_001191333.1
27, 28
3377
NM_001320874.1
NP_001307803.1
26, 27
3391
NM_020987.4
NP_066267.2
26, 27
3391
XM_005269715.3
XP_005269772.1
27, 28
3278
XM_006717796.3
XP_006717859.1
28, 29
3290
XM_006717802.3
XP_006717865.1
28, 29
3290
XM_011539708.2
XP_011538010.1
28, 29
3290
XM_011539709.2
XP_011538011.1
28, 29
3290
XM_017016110.1
XP_016871599.1
28, 29
3290
XM_017016111.1
XP_016871600.1
27, 28
3278
XM_017016112.1
XP_016871601.1
28, 29
3275
XM_017016113.1
XP_016871602.1
28, 29
3290
XM_017016114.1
XP_016871603.1
28, 29
3389
XM_017016115.1
XP_016871604.1
28, 29
3119
XM_017016116.1
XP_016871605.1
28, 29
3290
XM_017016117.1
XP_016871606.1
28, 29
3290
XM_017016118.1
XP_016871607.1
28, 29
3290
XM_017016119.1
XP_016871608.1
28, 29
3290
XM_017016120.1
XP_016871609.1
28, 29
3290
XM_017016121.1
XP_016871610.1
28, 29
3290
XM_017016122.1
XP_016871611.1
26, 27
3224
XM_017016123.1
XP_016871612.1
28, 29
3290
XM_017016124.1
XP_016871613.1
28, 29
3290
XM_017016125.1
XP_016871614.1
27, 28
3263
XM_017016126.1
XP_016871615.1
26, 27
3158
XM_017016127.1
XP_016871616.1
26, 27
3158
XM_017016128.1
XP_016871617.1
28, 29
3290
XM_017016129.1
XP_016871618.1
28, 29
3290
XM_017016130.1
XP_016871619.1
26, 27
3224
XM_017016131.1
XP_016871620.1
26, 27
3209
XM_017016132.1
XP_016871621.1
26, 27
3224
XM_017016133.1
XP_016871622.1
26, 27
3323
XM_017016134.1
XP_016871623.1
26, 27
3158
XM_017016135.1
XP_016871624.1
26, 27
3323
XM_017016136.1
XP_016871625.1
28, 29
3290
XM_017016137.1
XP_016871626.1
28, 29
3290
XM_017016138.1
XP_016871627.1
28, 29
3290
XM_017016139.1
XP_016871628.1
26, 27
3224
XM_017016140.1
XP_016871629.1
26, 27
3224
XM_017016141.1
XP_016871630.1
26, 27
3158
XM_017016142.1
XP_016871631.1
25, 26
3146
XM_017016143.1
XP_016871632.1
26, 27
3158
XM_017016144.1
XP_016871633.1
26, 27
3158
XM_017016145.1
XP_016871634.1
25, 26
3146
XM_017016146.1
XP_016871635.1
25, 26
3146
XM_017016147.1
XP_016871636.1
25, 26
3146
XM_017016148.1
XP_016871637.1
26, 27
3040
XM_017016149.1
XP_016871638.1
25, 26
3245
XM_017016150.1
XP_016871639.1
26, 27
3224
GenBank mRNA
AK092527.1
-
4, 5
1001
AK126851.1
-
8, 9
1079
AK292927.1
-
4, 5
882
AK295661.1
-
27, 28
3084
AL136710.1
-
4, 5
655
BX537917.1
-
27, 28
3005
BX648574.1
-
27, 28
3099
U13616.1
-
26, 27
3137
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
mitotic cytokinesis
ER to Golgi vesicle-mediated transport
plasma membrane organization
cytoskeletal anchoring at plasma membrane
signal transduction
axonogenesis
axon guidance
neuromuscular junction development
positive regulation of gene expression
positive regulation of cell communication by electrical coupling
positive regulation of sodium ion transport
magnesium ion homeostasis
neuronal action potential
positive regulation of homotypic cell-cell adhesion
Golgi to plasma membrane protein transport
regulation of potassium ion transport
establishment of protein localization
positive regulation of membrane potential
cellular response to magnesium ion
membrane assembly
protein localization to plasma membrane
maintenance of protein location in plasma membrane
protein targeting to plasma membrane
positive regulation of protein targeting to membrane
positive regulation of membrane depolarization during cardiac muscle cell action potential
negative regulation of delayed rectifier potassium channel activity
positive regulation of sodium ion transmembrane transporter activity
positive regulation of cation channel activity
structural constituent of cytoskeleton
protein binding
cytoskeletal protein binding
cytoskeletal adaptor activity
spectrin binding
protein binding, bridging
ion channel binding
cadherin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0