Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000043.5 | NP_000034.1 | 9 | 1120 | ||
NM_001320619.1 | NP_001307548.1 | 8 | 1037 | |||
NM_152871.3 | NP_690610.1 | 8 | 1057 | |||
NM_152872.3 | NP_690611.1 | 8 | 1095 | |||
NR_028033.3 | 7 | 948 | ||||
NR_028034.3 | 6 | 810 | ||||
NR_028035.3 | 7 | 873 | ||||
NR_028036.3 | 8 | 1011 | ||||
NR_135313.1 | 7 | 928 | ||||
NR_135314.1 | 9 | 1111 | ||||
NR_135315.1 | 7 | 864 | ||||
XM_006717819.3 | XP_006717882.1 | 10 | 5802 | |||
XM_011539764.2 | XP_011538066.1 | 10 | 1191 | |||
XM_011539765.2 | XP_011538067.1 | 9 | 916 | |||
XM_011539766.2 | XP_011538068.1 | 10 | 931 | |||
XM_011539767.2 | XP_011538069.1 | 9 | 854 | |||
GenBank mRNA | AB209361.1 | - | 10 | 4437 | ||
AK290978.1 | - | 9 | 1048 | |||
AY495076.2 | - | 9 | 996 | |||
BC012479.1 | - | 9 | 1112 | 4514272 | ||
CR450307.1 | - | 9 | 802 | |||
EU481974.2 | - | 9 | 997 | |||
FJ200481.1 | - | 8 | 719 | |||
FJ373045.1 | - | 9 | 803 | |||
FJ373046.1 | - | 9 | 801 | |||
FM246458.1 | - | 9 | 802 | |||
FM246459.1 | - | 9 | 803 | |||
M67454.1 | - | 9 | 996 | |||
X63717.1 | - | 9 | 1022 | |||
X83490.1 | - | 7 | 580 | |||
X83491.1 | - | 6 | 517 | |||
X83492.1 | - | 7 | 635 | |||
X83493.1 | - | 8 | 764 | |||
Z47993.1 | - | 8 | 739 | |||
Z47994.1 | - | 7 | 555 | |||
Z47995.1 | - | 6 | 492 | |||
Z66556.1 | - | 8 | 777 | |||
Z70519.1 | - | 8 | 693 | |||
Z70520.1 | - | 7 | 630 |