Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256196.1 | NP_001243125.1 | 11 | 1356 | ||
NM_001654.4 | NP_001645.1 | 11 | 1347 | |||
XM_006724529.2 | XP_006724592.1 | 11 | 1283 | |||
XM_011543906.2 | XP_011542208.1 | 11 | 2673 | |||
XM_011543908.2 | XP_011542210.1 | 11 | 2657 | |||
XM_011543909.2 | XP_011542211.1 | 6 | 648 | |||
GenBank mRNA | AB208831.1 | - | 9 | 3817 | ||
AF052136.1 | - | 1 | 54 | |||
AK130043.1 | - | 11 | 1240 | |||
AK297453.1 | - | 9 | 1070 | |||
AK300975.1 | - | 11 | 1203 | |||
BC002466.2 | - | 11 | 1227 | 3347509 | ||
BC007514.2 | - | 11 | 1234 | 2966604 | ||
BT019864.1 | - | 10 | 1136 | |||
KU177914.1 | - | 10 | 1136 | |||
KU177915.1 | - | 10 | 1151 | |||
X04790.1 | - | 11 | 1330 |