Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARSE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000047.2 | NP_000038.2 | 3 | 367 | ||
NM_001282628.1 | NP_001269557.1 | 4 | 653 | |||
XM_005274518.2 | XP_005274575.1 | 3 | 294 | |||
XM_005274519.4 | XP_005274576.1 | 3 | 457 | |||
XM_011545521.1 | XP_011543823.1 | 3 | 498 | |||
XM_017029525.1 | XP_016885014.1 | 4 | 584 | |||
XM_017029526.1 | XP_016885015.1 | 4 | 698 | |||
GenBank mRNA | AK223183.1 | - | 3 | 197 | ||
AK223199.1 | - | 3 | 164 | |||
AK301004.1 | - | 4 | 484 | |||
AK313093.1 | - | 3 | 186 | |||
BC130438.1 | - | 3 | 162 | 40146469 | ||
DA915010.1 | - | 3 | 369 | |||
DB073314.1 | - | 3 | 349 | |||
HX786884.1 | - | 3 | 258 | |||
X83573.1 | - | 3 | 184 |