Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001695.4 | NP_001686.1 | 5 | 575 | ||
GenBank mRNA | AK000064.1 | - | 5 | 503 | ||
AK298617.1 | - | 3 | 250 | |||
AK308632.1 | - | 4 | 336 | |||
AL707882.1 | - | 5 | 580 | |||
AU137773.2 | - | 5 | 519 | |||
BC010960.1 | - | 5 | 501 | 4108687 | ||
BX648079.1 | - | 5 | 492 | |||
J05682.1 | - | 4 | 225 | |||
X69151.1 | - | 5 | 496 |