Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127610.1 | NP_001121082.1 | 4 | 809 | ||
NM_001701.3 | NP_001692.1 | 4 | 910 | |||
GenBank mRNA | AK315347.1 | - | 4 | 854 | ||
BC009567.1 | - | 4 | 910 | 4071788 | ||
BC107424.1 | - | 4 | 914 | 4766834 | ||
CR541918.1 | - | 3 | 700 | |||
L34081.1 | - | 4 | 884 |