Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKX3-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001189.3 | NP_001180.1 | 1 | 131 | ||
GenBank mRNA | AF009801.1 | - | 1 | 55 | ||
BC111966.1 | - | 1 | 131 | 8327434 |