Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252678.1 | NP_001239607.1 | 11 | 959 | ||
NM_001320665.1 | NP_001307594.1 | 11 | 937 | |||
NM_033028.4 | NP_149017.2 | 12 | 1006 | |||
NR_045565.1 | 13 | 1064 | ||||
NR_045566.1 | 12 | 1319 | ||||
XM_011521848.1 | XP_011520150.1 | 12 | 981 | |||
XM_011521849.1 | XP_011520151.1 | 11 | 869 | |||
XM_011521851.1 | XP_011520153.1 | 10 | 857 | |||
XM_017022450.1 | XP_016877939.1 | 11 | 1218 | |||
XM_017022451.1 | XP_016877940.1 | 11 | 947 | |||
XM_017022452.1 | XP_016877941.1 | 11 | 1177 | |||
XM_017022453.1 | XP_016877942.1 | 11 | 864 | |||
XM_017022454.1 | XP_016877943.1 | 10 | 814 | |||
GenBank mRNA | AF359281.1 | - | 12 | 978 | ||
AK075321.1 | - | 12 | 970 | |||
AK298922.1 | - | 13 | 1036 | |||
AK302224.1 | - | 12 | 1273 | |||
AK303706.1 | - | 11 | 954 | |||
AK310106.1 | - | 4 | 393 | |||
AK314507.1 | - | 12 | 978 | |||
AY457143.1 | - | 12 | 1078 | |||
BC008923.2 | - | 9 | 783 | |||
BC027624.2 | - | 12 | 976 | 4824276 | ||
BX647855.1 | - | 11 | 930 | |||
KU177936.1 | - | 12 | 957 | |||
KU177937.1 | - | 3 | 270 |