Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004326.3 | NP_004317.2 | 8 | 2804 | ||
XM_005272971.4 | XP_005273028.1 | 8 | 2809 | |||
XM_005277417.3 | XP_005277474.1 | 8 | 2833 | |||
XM_005277418.3 | XP_005277475.1 | 8 | 2689 | |||
XM_017002026.1 | XP_016857515.1 | 8 | 2587 | |||
GenBank mRNA | BC116451.1 | - | 5 | 2199 | 7961941 | |
CN281037.1 | - | 1 | 320 | |||
Y13620.1 | - | 8 | 2892 |