Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001734.3 | NP_001725.1 | 8 | 1245 | ||
NM_201442.2 | NP_958850.1 | 8 | 1332 | |||
XM_005253760.1 | XP_005253817.1 | 8 | 1224 | |||
GenBank mRNA | AK025309.1 | - | 5 | 720 | ||
AK290295.1 | - | 8 | 1279 | |||
AM886411.1 | - | 7 | 967 | |||
BC056903.1 | - | 8 | 1272 | 6199186 | ||
J04080.1 | - | 8 | 1171 | |||
M18767.1 | - | 8 | 1191 |