Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM50B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006134.6 | NP_006125.2 | 5 | 531 | ||
NR_040016.1 | 5 | 531 | ||||
XM_011529746.1 | XP_011528048.1 | 5 | 529 | |||
GenBank mRNA | AF045606.2 | - | 5 | 476 | ||
AF086280.1 | - | 4 | 365 | |||
AK309977.1 | - | 5 | 458 | |||
AK310701.1 | - | 5 | 387 | |||
AK311870.1 | - | 4 | 351 | |||
AY358634.1 | - | 5 | 455 | |||
AY820138.1 | - | 4 | 318 | |||
BC000569.2 | - | 5 | 447 | 3342648 | ||
DQ286229.1 | - | 4 | 318 |