Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323374.1 | NP_001310303.1 | 10 | 906 | ||
NM_001323375.1 | NP_001310304.1 | 10 | 938 | |||
NM_001323376.1 | NP_001310305.1 | 7 | 908 | |||
NM_001323377.1 | NP_001310306.1 | 8 | 666 | |||
NM_001744.5 | NP_001735.1 | 9 | 1054 | |||
GenBank mRNA | AK311662.1 | - | 7 | 763 | ||
AK312840.1 | - | 9 | 909 | |||
BC016695.1 | - | 9 | 898 | |||
BC025687.1 | - | 9 | 896 | 5205478 | ||
D30742.1 | - | 9 | 884 | |||
L17000.1 | - | 9 | 848 | |||
L24959.1 | - | 10 | 906 |