Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146068.1 | NP_001139540.1 | 5 | 641 | ||
NM_001748.4 | NP_001739.2 | 5 | 863 | |||
XM_011544286.2 | XP_011542588.1 | 5 | 1040 | |||
GenBank mRNA | AF261089.1 | - | 5 | 766 | ||
AK297800.1 | - | 5 | 641 | |||
AK315179.1 | - | 5 | 765 | |||
AK316211.1 | - | 5 | 684 | |||
BC007686.2 | - | 5 | 863 | |||
BC011828.2 | - | 5 | 863 | |||
BC021303.2 | - | 5 | 757 | 5090865 | ||
DA448245.1 | - | 2 | 222 | |||
M23254.1 | - | 5 | 769 |