Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAST SIRNA Assays ›
Gene Symbol:
CAST
Gene Name:
calpastatin
Gene Aliases:
BS-17, PLACK
Chromosome Location:
Chr.5: 96662037 - 96774683 on Build GRCh38
UniGene ID:
Hs.436186
Species:
Human
siRNA ID
HSS101362
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAST
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042440.3
NP_001035905.1
17
1549
NM_001190442.1
NP_001177371.1
18
1421
NM_001284212.2
NP_001271141.1
15
1200
NM_001284213.2
NP_001271142.1
14
1107
NM_173060.3
NP_775083.1
16
1257
NR_104285.1
4
413
XM_006714696.3
XP_006714759.1
20
1563
XM_006714697.3
XP_006714760.1
19
1580
XM_006714698.3
XP_006714761.1
18
1541
XM_006714699.3
XP_006714762.1
19
1504
XM_006714700.3
XP_006714763.1
18
1523
XM_006714701.3
XP_006714764.1
18
1539
XM_006714702.3
XP_006714765.1
18
1415
XM_006714703.3
XP_006714766.1
17
1500
XM_006714704.3
XP_006714767.1
18
1448
XM_006714705.3
XP_006714768.1
17
1457
XM_006714706.3
XP_006714769.1
17
1375
XM_006714707.3
XP_006714770.1
17
1326
XM_006714708.3
XP_006714771.1
16
1286
XM_006714709.3
XP_006714772.1
16
1249
XM_006714710.3
XP_006714773.1
15
1204
XM_006714711.3
XP_006714774.1
15
1249
XM_006714712.3
XP_006714775.1
14
1196
XM_006714713.3
XP_006714776.1
4
413
XM_006714714.3
XP_006714777.1
4
413
XM_006714715.3
XP_006714778.1
5
1139
XM_011543654.2
XP_011541956.1
18
1499
XM_011543655.2
XP_011541957.1
18
1496
XM_011543656.2
XP_011541958.1
17
1443
XM_011543657.2
XP_011541959.1
17
1325
XM_011543658.2
XP_011541960.1
15
1178
XM_017009911.1
XP_016865400.1
18
1541
XM_017009912.1
XP_016865401.1
19
1552
XM_017009913.1
XP_016865402.1
19
1521
XM_017009914.1
XP_016865403.1
18
1523
XM_017009915.1
XP_016865404.1
19
1525
XM_017009916.1
XP_016865405.1
18
1539
XM_017009917.1
XP_016865406.1
18
1457
XM_017009918.1
XP_016865407.1
17
1484
XM_017009919.1
XP_016865408.1
17
1457
XM_017009920.1
XP_016865409.1
17
1367
XM_017009921.1
XP_016865410.1
17
1384
XM_017009922.1
XP_016865411.1
16
1418
XM_017009923.1
XP_016865412.1
16
1418
XM_017009924.1
XP_016865413.1
16
1400
XM_017009925.1
XP_016865414.1
16
1336
XM_017009926.1
XP_016865415.1
16
1336
XM_017009927.1
XP_016865416.1
17
1325
XM_017009928.1
XP_016865417.1
16
1324
XM_017009929.1
XP_016865418.1
16
1243
XM_017009930.1
XP_016865419.1
15
1210
XM_017009931.1
XP_016865420.1
15
1204
XM_017009932.1
XP_016865421.1
14
1267
GenBank mRNA
AB208816.1
-
18
1385
AF327443.1
-
13
1099
AK292289.1
-
4
413
AK292356.1
-
4
386
AK296919.1
-
18
1379
AK297879.1
-
16
1199
AK298541.1
-
17
1326
AK299402.1
-
17
1306
AK300624.1
-
14
1090
AK302206.1
-
5
413
AK303836.1
-
10
780
AK311585.1
-
5
591
AK311598.1
-
5
467
AK314290.1
-
15
1202
AK315146.1
-
15
1201
AK315673.1
-
4
361
AL832174.1
-
16
1242
AL832349.1
-
16
1245
AY189741.1
-
6
412
BC013579.1
-
17
1252
3878564
BC078149.1
-
12
835
BT009783.1
-
14
1034
BX648397.1
-
15
1151
D16217.1
-
17
1319
DB033043.1
-
4
361
U26724.2
-
8
682
U31345.1
-
6
429
U58996.2
-
4
392
Back To Top
More Information
Panther Classification:
Molecular Function -
protein-binding activity modulator
protease inhibitor
Gene Ontology Categories:
Function(s)
Process(es)
liver development
egg activation
brain development
myoblast fusion
aging
negative regulation of endopeptidase activity
organ regeneration
myoblast differentiation
negative regulation of cell cycle arrest
cell-cell adhesion
negative regulation of type B pancreatic cell apoptotic process
protease binding
endopeptidase inhibitor activity
protein binding
calcium-dependent cysteine-type endopeptidase inhibitor activity
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0