Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014437.2 | NP_001014437.1 | 16 | 1980 | ||
NM_001194997.1 | NP_001181926.1 | 16 | 1980 | |||
NM_001751.5 | NP_001742.1 | 15 | 1731 | |||
NM_139273.3 | NP_644802.1 | 15 | 1731 | |||
NR_036542.1 | 16 | 2027 | ||||
XM_006718341.3 | XP_006718404.1 | 15 | 1754 | |||
XM_017018389.1 | XP_016873878.1 | 15 | 1754 | |||
XM_017018390.1 | XP_016873879.1 | 16 | 1994 | |||
XM_017018391.1 | XP_016873880.1 | 13 | 1978 | |||
XM_017018392.1 | XP_016873881.1 | 15 | 1916 | |||
GenBank mRNA | AB208814.1 | - | 12 | 4948 | ||
AF288206.1 | - | 15 | 1664 | |||
AF288207.1 | - | 15 | 1664 | |||
AK096313.1 | - | 16 | 1994 | |||
AK222614.1 | - | 15 | 1672 | |||
AK289432.1 | - | 15 | 1704 | |||
AK295371.1 | - | 15 | 1622 | |||
AK296763.1 | - | 16 | 1845 | |||
AK298514.1 | - | 16 | 1947 | |||
AK302644.1 | - | 16 | 1947 | |||
AK308005.1 | - | 16 | 1898 | |||
BC002880.2 | - | 15 | 1717 | 3940167 | ||
BT009913.1 | - | 15 | 1647 | |||
BX647906.1 | - | 16 | 1954 | |||
L06845.1 | - | 13 | 1518 |