Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319145.1 | NP_001306074.1 | 3 | 790 | ||
NM_001766.3 | NP_001757.1 | 3 | 790 | |||
XM_005245583.1 | XP_005245640.1 | 2 | 469 | |||
XM_006711621.1 | XP_006711684.1 | 2 | 351 | |||
XM_011510125.1 | XP_011508427.1 | 3 | 605 | |||
XM_011510127.1 | XP_011508429.1 | 2 | 433 | |||
GenBank mRNA | AK225996.1 | - | 2 | 504 | ||
AK310541.1 | - | 2 | 351 | |||
BC027926.1 | - | 3 | 790 | 5229534 | ||
J04142.1 | - | 2 | 455 |