Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK223393.1 | - | 3 | 601 | ||
AK301154.1 | - | 3 | 624 | |||
AK310676.1 | - | 3 | 463 | |||
AK313614.1 | - | 3 | 630 | |||
BC033583.1 | - | 3 | 585 | 5227138 | ||
CR996443.1 | - | 3 | 607 | |||
M14362.1 | - | 3 | 581 | |||
M16336.1 | - | 3 | 595 | |||
M16445.1 | - | 3 | 578 |