Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000073.2 | NP_000064.1 | 4 | 444 | ||
XM_005271724.3 | XP_005271781.1 | 4 | 481 | |||
XM_006718941.2 | XP_006719004.1 | 4 | 432 | |||
GenBank mRNA | AK223314.1 | - | 4 | 397 | ||
AK310578.1 | - | 5 | 572 | |||
AK310622.1 | - | 5 | 602 | |||
AK313966.1 | - | 4 | 432 | |||
AK315895.1 | - | 4 | 388 | |||
BC113830.1 | - | 4 | 390 | 40029587 | ||
X04145.1 | - | 4 | 401 |