Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031666.1 | NP_001026836.1 | 4 | 401 | ||
NM_001031809.1 | NP_001026979.1 | 5 | 572 | |||
NM_006138.4 | NP_006129.4 | 6 | 710 | |||
XM_011545363.2 | XP_011543665.1 | 5 | 457 | |||
GenBank mRNA | AY095480.1 | - | 4 | 444 | ||
BC008487.1 | - | 6 | 655 | 4102067 | ||
BE564041.1 | - | 4 | 422 | |||
BF210971.1 | - | 444 | ||||
L35848.1 | - | 6 | 680 |