Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000072.3 | NP_000063.2 | 5 | 773 | ||
NM_001001547.2 | NP_001001547.1 | 5 | 734 | |||
NM_001001548.2 | NP_001001548.1 | 5 | 773 | |||
NM_001127443.1 | NP_001120915.1 | 4 | 479 | |||
NM_001127444.1 | NP_001120916.1 | 3 | 654 | |||
NM_001289908.1 | NP_001276837.1 | 3 | 447 | |||
NM_001289909.1 | NP_001276838.1 | 4 | 538 | |||
NM_001289911.1 | NP_001276840.1 | 3 | 412 | |||
NR_110501.1 | 4 | 538 | ||||
XM_005250713.1 | XP_005250770.1 | 5 | 715 | |||
XM_005250714.1 | XP_005250771.1 | 5 | 682 | |||
XM_005250715.4 | XP_005250772.1 | 5 | 840 | |||
GenBank mRNA | AK096858.1 | - | 8 | 1043 | ||
AK129899.1 | - | 5 | 715 | |||
AK300101.1 | - | 3 | 412 | |||
AK312920.1 | - | 5 | 697 | |||
BC008406.1 | - | 5 | 648 | 4244251 | ||
DA566475.1 | - | 4 | 479 | |||
HM217020.1 | - | 3 | 456 | |||
HM217021.1 | - | 3 | 456 | |||
HM217022.1 | - | 3 | 456 | |||
HM217023.1 | - | 3 | 456 | |||
HM217024.1 | - | 3 | 456 | |||
HM217025.1 | - | 3 | 456 | |||
HM217026.1 | - | 3 | 447 | |||
KU177961.1 | - | 3 | 359 | |||
L06850.1 | - | 4 | 491 | |||
M24795.1 | - | 5 | 569 | |||
M98398.1 | - | 5 | 613 | |||
M98399.1 | - | 5 | 613 | |||
S67532.1 | - | 4 | 495 |