Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ENTPD6 SIRNA Assays ›
Gene Symbol:
ENTPD6
Gene Name:
ectonucleoside triphosphate diphosphohydrolase 6 (putative)
Gene Aliases:
CD39L2, IL-6SAG, IL6ST2, NTPDase-6, dJ738P15.3
Chromosome Location:
Chr.20: 25195670 - 25228075 on Build GRCh38
UniGene ID:
Hs.500375
Species:
Human
siRNA ID
HSS101586
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ENTPD6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001114089.3
NP_001107561.1
4
622
NM_001247.4
NP_001238.2
5
691
NM_001317941.2
NP_001304870.1
5
688
NM_001322378.1
NP_001309307.1
5
835
NM_001322379.1
NP_001309308.1
3
300
NM_001322380.1
NP_001309309.1
3
300
NM_001322383.1
NP_001309312.1
6
879
NM_001322384.1
NP_001309313.1
3
300
NM_001322386.1
NP_001309315.1
4
622
NM_001322388.1
NP_001309317.1
5
691
NM_001322389.1
NP_001309318.1
4
766
NM_001322390.1
NP_001309319.1
6
813
NM_001322391.1
NP_001309320.1
5
691
NM_001322392.1
NP_001309321.1
4
619
NM_001322393.1
NP_001309322.1
4
622
NM_001322394.1
NP_001309323.1
4
619
NM_001322395.1
NP_001309324.1
4
766
NM_001322396.1
NP_001309325.1
5
688
NM_001322397.1
NP_001309326.1
5
691
NM_001322398.1
NP_001309327.1
4
766
XM_006723665.2
XP_006723728.1
6
727
XM_011529396.1
XP_011527698.1
6
731
XM_011529397.1
XP_011527699.1
6
660
XM_017028129.1
XP_016883618.1
4
500
XM_017028131.1
XP_016883620.1
4
500
XM_017028133.1
XP_016883622.1
4
470
XM_017028134.1
XP_016883623.1
3
267
GenBank mRNA
AF039916.1
-
5
625
AK074691.1
-
4
471
AK128412.1
-
3
239
AK293256.1
-
4
563
AK297954.1
-
5
531
AK300028.1
-
3
251
AK302395.1
-
5
539
AK303786.1
-
4
577
AK308626.1
-
4
589
AL834158.1
-
4
537
AY327581.1
-
5
588
BC025980.2
-
5
595
3636163
CR936750.1
-
3
248
CR936765.1
-
5
605
Back To Top
More Information
Panther Classification:
Molecular Function -
hydrolase
nucleotide phosphatase
phosphatase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
response to magnesium ion
response to calcium ion
guanosine-5'-triphosphate,3'-diphosphate diphosphatase activity
nucleoside-triphosphatase activity
uridine-diphosphatase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0