Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297649.1 | NP_001284578.1 | 7, 8 | 762 | ||
NM_004356.3 | NP_004347.1 | 7, 8 | 880 | |||
XM_011520492.2 | XP_011518794.1 | 7, 8 | 1114 | |||
XM_017018598.1 | XP_016874087.1 | 7, 8 | 623 | |||
GenBank mRNA | BC002978.1 | - | 7, 8 | 687 | 3510634 | |
BC093047.1 | - | 7, 8 | 878 | 30398483 | ||
BT019507.1 | - | 7, 8 | 647 | |||
BT019508.1 | - | 7, 8 | 647 | |||
M33680.1 | - | 7, 8 | 885 |