Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001220488.1 | NP_001207417.1 | 11, 12 | 1933 | ||
NM_001220489.1 | NP_001207418.1 | 9, 10 | 1710 | |||
NM_001220490.1 | NP_001207419.1 | 9, 10 | 1618 | |||
NM_001257.4 | NP_001248.1 | 10, 11 | 1827 | |||
XM_011522804.2 | XP_011521106.1 | 9, 10 | 1434 | |||
GenBank mRNA | AK294277.1 | - | 4, 5 | 690 | ||
AK295886.1 | - | 9, 10 | 1448 | |||
AK298612.1 | - | 9, 10 | 1536 | |||
AK304592.1 | - | 3, 4 | 326 | |||
AK304733.1 | - | 11, 12 | 1688 | |||
BC028624.2 | - | 10, 11 | 1619 | 4825734 | ||
BC030653.2 | - | 10, 11 | 1619 | 4811561 | ||
CA445317.1 | - | 4, 5 | 677 | |||
DA561809.1 | - | 3, 4 | 503 | |||
L34058.1 | - | 10, 11 | 1972 | |||
U59288.1 | - | 10, 11 | 1744 |