Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001220488.1 | NP_001207417.1 | 6 | 1024 | ||
NM_001220489.1 | NP_001207418.1 | 4 | 801 | |||
NM_001220490.1 | NP_001207419.1 | 4 | 709 | |||
NM_001257.4 | NP_001248.1 | 5 | 918 | |||
XM_011522804.2 | XP_011521106.1 | 4 | 525 | |||
XM_017022848.1 | XP_016878337.1 | 6 | 941 | |||
GenBank mRNA | AK295886.1 | - | 4 | 539 | ||
AK298612.1 | - | 4 | 627 | |||
AK304733.1 | - | 6 | 779 | |||
AK307862.1 | - | 6 | 715 | |||
BC028624.2 | - | 5 | 710 | 4825734 | ||
BC030653.2 | - | 5 | 710 | 4811561 | ||
BI756586.1 | - | 5 | 722 | |||
BU162665.1 | - | 1 | 51 | |||
L34058.1 | - | 5 | 1063 | |||
U59288.1 | - | 5 | 835 |