Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144663.1 | NP_001138135.1 | 12 | 1659 | ||
NM_004063.3 | NP_004054.3 | 12 | 1658 | |||
XM_011516790.1 | XP_011515092.1 | 12 | 1657 | |||
GenBank mRNA | AK300638.1 | - | 4 | 537 | ||
AK301001.1 | - | 8 | 984 | |||
AK312873.1 | - | 12 | 1659 | |||
BC112013.1 | - | 12 | 1631 | 8327481 | ||
BC113464.1 | - | 12 | 1619 | 8322516 | ||
U07969.1 | - | 12 | 1620 | |||
X83228.1 | - | 12 | 1625 |