Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145306.1 | NP_001138778.1 | 7, 8 | 1344 | ||
NM_001259.6 | NP_001250.1 | 7, 8 | 1239 | |||
XM_006715835.2 | XP_006715898.1 | 7, 8 | 1464 | |||
GenBank mRNA | AK313491.1 | - | 7, 8 | 1282 | ||
BC052264.1 | - | 7, 8 | 1220 | 6377987 | ||
X66365.1 | - | 6, 7 | 944 |