Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEBPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285829.1 | NP_001272758.1 | 1 | 1203 | ||
NM_001287424.1 | NP_001274353.1 | 1 | 1203 | |||
NM_001287435.1 | NP_001274364.1 | 1 | 1203 | |||
NM_004364.4 | NP_004355.2 | 1 | 1203 | |||
GenBank mRNA | BC063874.1 | - | 2 | 732 | ||
X87248.1 | - | 1 | 1085 | |||
Y11525.1 | - | 1 | 1199 |