Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267.2 | NP_001258.2 | 1 | 769 | ||
XM_011524214.2 | XP_011522516.1 | 1 | 1784 | |||
GenBank mRNA | AF371328.1 | - | 1 | 782 | ||
AK292177.1 | - | 1 | 812 | |||
BC036360.1 | - | 1 | 767 | 5163623 | ||
BC073974.1 | - | 1 | 769 | 5730623 |