Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256125.1 | NP_001243054.2 | 8, 9 | 1074 | ||
NM_003465.2 | NP_003456.1 | 9, 10 | 1131 | |||
NR_045785.1 | 9, 10 | 1131 | ||||
GenBank mRNA | AK055165.1 | - | 8, 9 | 1014 | ||
AK131093.1 | - | 10, 11 | 5266 | |||
AK314331.1 | - | 9, 10 | 1070 | |||
BC069614.1 | - | 8, 9 | 1000 | |||
BC103695.1 | - | 9, 10 | 1047 | 40005693 | ||
BC105680.1 | - | 9, 10 | 1047 | 40005699 | ||
BC105681.1 | - | 9, 10 | 1047 | 40005697 | ||
BC105682.1 | - | 9, 10 | 1047 | 40005696 | ||
U29615.1 | - | 9, 10 | 1047 | |||
U62662.1 | - | 9, 10 | 1044 |