Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256944.1 | NP_001243873.1 | 9 | 2137 | ||
NM_001830.3 | NP_001821.2 | 11 | 2392 | |||
GenBank mRNA | AB019432.1 | - | 10 | 2237 | ||
AF170492.1 | - | 9 | 1962 | |||
AK225287.1 | - | 11 | 2353 | |||
AK289564.1 | - | 11 | 2339 | |||
AK299611.1 | - | 8 | 1770 | |||
BC036068.1 | - | 10 | 2012 | 5311366 | ||
BC130278.1 | - | 10 | 2251 | 40146309 | ||
X77197.1 | - | 11 | 2345 |