Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000090.3 | NP_000081.1 | 17 | 1287 | ||
GenBank mRNA | BC028178.1 | - | 17 | 1285 | 5405119 | |
S79877.1 | - | 4 | 169 | |||
X14420.1 | - | 17 | 1272 | |||
X15332.1 | - | 13 | 729 |