Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000090.3 | NP_000081.1 | 1 | 163 | ||
GenBank mRNA | AK308010.1 | - | 1 | 163 | ||
BC028178.1 | - | 1 | 161 | 5405119 | ||
BP374999.1 | - | 1 | 163 | |||
X07240.1 | - | 1 | 46 | |||
X14420.1 | - | 1 | 148 |