Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL6A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001848.2 | NP_001839.2 | 35 | 2819 | ||
GenBank mRNA | AK298968.1 | - | 18 | 1527 | ||
BC005159.2 | - | 9 | 969 | |||
BC022236.1 | - | 6 | 951 | |||
BC052575.1 | - | 35 | 2799 | 6598940 | ||
X15880.1 | - | 8 | 941 |