Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001869.2 | NP_001860.2 | 1 | 70 | ||
GenBank mRNA | AK293378.1 | - | 1 | 66 | ||
BC007009.1 | - | 1 | 70 | 3949984 | ||
BC014571.1 | - | 1 | 72 | 3950117 | ||
BC015140.1 | - | 1 | 72 | 4040975 | ||
BT007403.1 | - | 1 | 45 | |||
KU177990.1 | - | 1 | 45 | |||
KU177991.1 | - | 1 | 45 | |||
KU177992.1 | - | 1 | 45 | |||
KU177993.1 | - | 1 | 45 | |||
U19977.1 | - | 1 | 49 |