Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000554.4 | NP_000545.1 | 4 | 615 | ||
GenBank mRNA | BC016664.1 | - | 4 | 550 | 3869862 | |
BC053672.1 | - | 4 | 501 | 6144191 | ||
BT007364.1 | - | 3 | 411 |