Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYBA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005208.4 | NP_005199.2 | 4 | 264 | ||
XM_017024198.1 | XP_016879687.1 | 5 | 560 | |||
GenBank mRNA | BC069537.1 | - | 4 | 257 | 7262224 | |
BC113693.1 | - | 4 | 258 | 8322745 | ||
KM230602.1 | - | 4 | 246 | |||
KM230603.1 | - | 4 | 245 | |||
KM230604.1 | - | 4 | 242 | |||
KM230605.1 | - | 3 | 242 | |||
KR336660.1 | - | 4 | 242 | |||
U59058.1 | - | 2 | 130 |