Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001888.4 | NP_001879.1 | 6, 7 | 840 | ||
GenBank mRNA | AK290852.1 | - | 4, 5 | 559 | ||
BC018061.1 | - | 4, 5 | 558 | 4817634 | ||
BI860684.1 | - | 5, 6 | 513 | |||
BX648477.1 | - | 6, 7 | 756 | |||
L02950.1 | - | 4, 5 | 519 | |||
U85772.1 | - | 4, 5 | 514 |