Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001315.2 | NP_001306.1 | 8 | 1136 | ||
NM_139012.2 | NP_620581.1 | 8 | 1136 | |||
NM_139013.2 | NP_620582.1 | 8 | 1136 | |||
NM_139014.2 | NP_620583.1 | 8 | 1136 | |||
XM_006714998.3 | XP_006715061.1 | 8 | 611 | |||
XM_011514310.2 | XP_011512612.2 | 8 | 1404 | |||
XM_017010299.1 | XP_016865788.1 | 8 | 1178 | |||
XM_017010300.1 | XP_016865789.1 | 8 | 1178 | |||
XM_017010301.1 | XP_016865790.1 | 8 | 1178 | |||
XM_017010302.1 | XP_016865791.1 | 8 | 1178 | |||
XM_017010303.1 | XP_016865792.1 | 8 | 1178 | |||
XM_017010304.1 | XP_016865793.1 | 8 | 1178 | |||
GenBank mRNA | AB074150.1 | - | 8 | 655 | ||
AF100544.1 | - | 8 | 670 | |||
AK291709.1 | - | 8 | 1031 | |||
AK303414.1 | - | 9 | 700 | |||
BC000092.1 | - | 8 | 993 | 3505234 | ||
BC031574.1 | - | 8 | 992 | 5181064 | ||
BT006933.1 | - | 8 | 655 | |||
CR536505.1 | - | 8 | 655 | |||
FJ032367.1 | - | 8 | 655 | |||
FJ032368.1 | - | 8 | 682 | |||
L35263.1 | - | 8 | 1017 | |||
L35264.1 | - | 8 | 863 |