Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317237.1 | NP_001304166.1 | 10 | 1102 | ||
NM_001908.4 | NP_001899.1 | 10 | 1133 | |||
NM_147780.3 | NP_680090.1 | 12 | 1295 | |||
NM_147781.3 | NP_680091.1 | 11 | 1252 | |||
NM_147782.3 | NP_680092.1 | 11 | 1221 | |||
NM_147783.3 | NP_680093.1 | 11 | 1207 | |||
XM_006716244.2 | XP_006716307.1 | 12 | 1319 | |||
XM_006716245.2 | XP_006716308.1 | 10 | 1423 | |||
XM_011543812.2 | XP_011542114.1 | 12 | 1228 | |||
XM_017013097.1 | XP_016868586.1 | 11 | 1209 | |||
XM_017013098.1 | XP_016868587.1 | 11 | 1209 | |||
XM_017013099.1 | XP_016868588.1 | 12 | 1251 | |||
XM_017013100.1 | XP_016868589.1 | 11 | 1153 | |||
XM_017013101.1 | XP_016868590.1 | 7 | 676 | |||
GenBank mRNA | AK075393.1 | - | 10 | 1109 | ||
AK092070.1 | - | 11 | 1144 | |||
AK097384.1 | - | 5 | 1063 | |||
AK128472.1 | - | 1 | 1726 | |||
AK130184.1 | - | 10 | 1070 | |||
AK290239.1 | - | 10 | 1070 | |||
AK296845.1 | - | 9 | 874 | |||
AK297687.1 | - | 8 | 834 | |||
BC010240.1 | - | 11 | 1150 | 4154307 | ||
BC095408.1 | - | 10 | 1070 | 30334082 | ||
BX647765.1 | - | 10 | 993 | |||
BX647952.1 | - | 10 | 1073 | |||
BX648239.1 | - | 5 | 6844 | |||
L16510.1 | - | 11 | 1135 | |||
M13230.1 | - | 6 | 568 |