Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201575.1 | NP_001188504.1 | 4 | 329 | ||
NM_001333.3 | NP_001324.2 | 4 | 532 | |||
GenBank mRNA | AB001928.1 | - | 4 | 355 | ||
AF070448.1 | - | 3 | 309 | |||
AK310645.1 | - | 4 | 801 | |||
AK310740.1 | - | 4 | 464 | |||
AK312806.1 | - | 4 | 329 | |||
AY358641.1 | - | 4 | 532 | |||
BC023504.2 | - | 4 | 346 | 3139314 | ||
BC110512.1 | - | 4 | 401 | 40031744 | ||
Y14734.1 | - | 4 | 351 |