Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000499.4 | NP_000490.1 | 2 | 902 | ||
NM_001319216.1 | NP_001306145.1 | 2 | 905 | |||
NM_001319217.1 | NP_001306146.1 | 2 | 905 | |||
GenBank mRNA | AK223108.1 | - | 2 | 823 | ||
AK223113.1 | - | 2 | 820 | |||
AK310810.1 | - | 2 | 949 | |||
AK313880.1 | - | 2 | 820 | |||
AM233517.1 | - | 2 | 194 | |||
AM233518.1 | - | 2 | 194 | |||
AY871801.1 | - | 2 | 789 | |||
BC023019.1 | - | 2 | 787 | 5123393 |