Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2C18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000772.2 | NP_000763.1 | 9 | 1748 | ||
NM_001128925.1 | NP_001122397.1 | 8 | 1571 | |||
GenBank mRNA | AK223510.1 | - | 9 | 1447 | ||
AK308558.1 | - | 9 | 1403 | |||
AK313403.1 | - | 9 | 1447 | |||
BC069666.1 | - | 9 | 1616 | 7262569 | ||
BC096257.1 | - | 9 | 1467 | 40006072 | ||
BC096258.1 | - | 9 | 1467 | 40006073 | ||
BC096260.1 | - | 8 | 1290 | 40006076 | ||
BX538127.1 | - | 6 | 882 | |||
M61853.1 | - | 9 | 1464 | |||
M61856.1 | - | 9 | 1621 |