Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP3A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000777.4 | NP_000768.1 | 9 | 965 | ||
NM_001291829.1 | NP_001278758.1 | 10 | 1465 | |||
NM_001291830.1 | NP_001278759.1 | 10 | 1225 | |||
NR_033807.2 | 9 | 2569 | ||||
XM_006715859.2 | XP_006715922.1 | 9 | 932 | |||
XM_011515843.1 | XP_011514145.1 | 10 | 1082 | |||
XM_017011781.1 | XP_016867270.1 | 6 | 607 | |||
XM_017011782.1 | XP_016867271.1 | 4 | 404 | |||
GenBank mRNA | AK055879.1 | - | 1 | 737 | ||
AK223008.1 | - | 9 | 935 | |||
AK296205.1 | - | 10 | 1192 | |||
AK313813.1 | - | 9 | 932 | |||
BC033862.1 | - | 9 | 924 | 5188554 | ||
BX537676.1 | - | 9 | 2540 | |||
BX648856.1 | - | 9 | 2521 | |||
J04813.1 | - | 9 | 922 | |||
L26985.1 | - | 11 | 1155 | |||
X90579.1 | - | 10 | 1054 |