Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGKQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001347.3 | NP_001338.2 | 18, 19 | 2298 | ||
XM_011513411.1 | XP_011511713.1 | 18, 19 | 2283 | |||
GenBank mRNA | AB209503.1 | - | 18, 19 | 2049 | ||
BC063801.1 | - | 18, 19 | 2306 | 6526640 | ||
GU727647.1 | - | 18, 19 | 2236 | |||
L38707.1 | - | 18, 19 | 2258 |