Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001917.4 | NP_001908.3 | 4, 5 | 542 | ||
XM_011538004.2 | XP_011536306.1 | 5, 6 | 628 | |||
XM_011538005.2 | XP_011536307.1 | 5, 6 | 632 | |||
GenBank mRNA | AK312995.1 | - | 4, 5 | 529 | ||
AX741655.1 | - | 4, 5 | 532 | |||
AX741656.1 | - | 4, 5 | 532 | |||
AX741658.1 | - | 3, 4 | 417 | |||
AX776478.1 | - | 4, 5 | 532 | |||
AX776479.1 | - | 4, 5 | 532 | |||
AX776480.1 | - | 7, 8 | 1544 | |||
AX786386.1 | - | 4, 5 | 532 | |||
BC029057.1 | - | 4, 5 | 512 | 5184628 | ||
BC074770.2 | - | 4, 5 | 414 | 30915193 | ||
BX538210.1 | - | 4, 5 | 474 | |||
KU178015.1 | - | 3, 4 | 389 | |||
KU178016.1 | - | 3, 4 | 389 |