Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000789.3 | NP_000780.1 | 19 | 2800 | ||
NM_001178057.1 | NP_001171528.1 | 8 | 1095 | |||
NM_152830.2 | NP_690043.1 | 8 | 1095 | |||
XM_006721737.3 | XP_006721800.2 | 8 | 1208 | |||
GenBank mRNA | AB208971.1 | - | 19 | 2790 | ||
AK301988.1 | - | 8 | 1066 | |||
J04144.1 | - | 19 | 2788 | |||
JQ927553.1 | - | 19 | 2800 | |||
M26657.1 | - | 8 | 1066 | |||
M29981.1 | - | 8 | 1144 | |||
X16295.1 | - | 8 | 1072 |