Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129889.2 | NP_001123361.1 | 9 | 1819 | ||
NM_001322182.1 | NP_001309111.1 | 6 | 942 | |||
NM_001322183.1 | NP_001309112.1 | 7 | 996 | |||
NM_001322184.1 | NP_001309113.1 | 8 | 1296 | |||
NM_001322185.1 | NP_001309114.1 | 7 | 1242 | |||
NM_001322186.1 | NP_001309115.1 | 9 | 1511 | |||
NM_001922.4 | NP_001913.2 | 7 | 1720 | |||
XM_011521049.1 | XP_011519351.1 | 9 | 2344 | |||
XM_017020401.1 | XP_016875890.1 | 9 | 1852 | |||
GenBank mRNA | AJ132932.1 | - | 1 | 114 | ||
AJ132933.1 | - | 1 | 114 | |||
BC028311.1 | - | 7 | 1567 | 4705935 | ||
D17547.1 | - | 7 | 1707 | |||
DQ902581.1 | - | 9 | 1684 | |||
L18967.1 | - | 7 | 1677 | |||
S69231.1 | - | 7 | 1642 |