Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001383.3 | NP_001374.3 | 7, 8 | 782 | ||
GenBank mRNA | AF321876.1 | - | 7, 8 | 782 | ||
AK090530.1 | - | 1, 2 | 855 | |||
AK297947.1 | - | 6, 7 | 800 | |||
AK314015.1 | - | 7, 8 | 768 | |||
BC003099.1 | - | 7, 8 | 744 | |||
BC096088.1 | - | 6, 7 | 638 | 40000799 | ||
BT019878.1 | - | 5, 6 | 524 | |||
S81752.1 | - | 7, 8 | 817 | |||
U34880.1 | - | 7, 8 | 817 |