Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000111.2 | NP_000102.1 | 2 | 254 | ||
XM_011515867.2 | XP_011514169.1 | 2 | 235 | |||
GenBank mRNA | BC025671.1 | - | 2 | 228 | 5202612 | |
BP262081.1 | - | 2 | 254 | |||
BX640837.1 | - | 2 | 247 | |||
L02785.1 | - | 2 | 227 |